Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146001.2 | NP_666113.2 | 11 | 1061 | ||
GenBank mRNA | AK087586.1 | - | 11 | 1059 | ||
AK147246.1 | - | 11 | 1017 | |||
AK147269.1 | - | 11 | 1017 | |||
AK166346.1 | - | 11 | 1030 | |||
BC017516.1 | - | 11 | 1054 | 4504441 |