Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146003.2 | NP_666115.2 | 18 | 3215 | ||
GenBank mRNA | AK028461.1 | - | 6 | 899 | ||
AK048485.1 | - | 13 | 2664 | |||
AK129219.1 | - | 14 | 2742 | |||
BC028838.1 | - | 1 | 331 | |||
BC052718.1 | - | 18 | 2982 | |||
BC061480.1 | - | 18 | 3038 | 6509771 | ||
BC151128.1 | - | 19 | 2646 | 9088041 | ||
BC172171.1 | - | 19 | 2646 | 9054841 |