Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 3 | 613 | ||
GenBank mRNA | AK032295.1 | - | 3 | 576 | ||
AK048357.1 | - | 3 | 510 | |||
AK156203.1 | - | 3 | 504 | |||
AK165939.1 | - | 3 | 473 | |||
AK178755.1 | - | 2 | 150 | |||
AK192551.1 | - | 1 | 128 | |||
AK197577.1 | - | 1 | 86 | |||
AK220105.1 | - | 2 | 150 | |||
BC026562.1 | - | 3 | 314 | 5028264 |