Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146057.3 | NP_666169.1 | 2 | 278 | ||
GenBank mRNA | AK014991.1 | - | 2 | 170 | ||
AK152216.1 | - | 2 | 245 | |||
AK152884.1 | - | 2 | 255 | |||
AK167579.1 | - | 2 | 202 | |||
AK167688.1 | - | 2 | 142 | |||
AK169162.1 | - | 2 | 210 | |||
AK171259.1 | - | 2 | 272 | |||
AK197429.1 | - | 1 | 36 | |||
BC010828.1 | - | 2 | 214 | 4220180 | ||
BC024876.1 | - | 2 | 242 | 3155209 | ||
BC057669.1 | - | 2 | 204 | 2655176 | ||
BY272917.1 | - | 2 | 279 |