Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146073.3 | NP_666185.3 | 5 | 1572 | ||
GenBank mRNA | AF542387.1 | - | 6 | 1241 | ||
AK041496.1 | - | 5 | 1255 | |||
AK041648.1 | - | 6 | 1568 | |||
AK046719.1 | - | 5 | 1560 | |||
AK080845.1 | - | 5 | 1576 | |||
BC059814.1 | - | 5 | 1615 | 6813525 | ||
BC086758.1 | - | 6 | 628 |