Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000655.4 | NP_000646.2 | 9 | 1572 | ||
NR_029467.1 | 7 | 1336 | ||||
GenBank mRNA | AJ246000.1 | - | 9 | 1443 | ||
AK225713.1 | - | 9 | 1483 | |||
AK298705.1 | - | 7 | 1331 | |||
CB164568.1 | - | 1 | 34 | |||
M25280.1 | - | 9 | 1483 | |||
X16070.1 | - | 9 | 1495 | |||
X16150.1 | - | 9 | 1459 | |||
X17519.1 | - | 9 | 1456 |