Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146096.3 | NP_666208.2 | 4 | 1145 | ||
GenBank mRNA | AK050879.1 | - | 4 | 1148 | ||
AK152807.1 | - | 2 | 404 | |||
BC032261.1 | - | 2 | 633 | |||
BC091733.1 | - | 3 | 542 |