Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146107.2 | NP_666219.1 | 4, 5 | 392 | ||
GenBank mRNA | AK078508.1 | - | 4, 5 | 408 | ||
AK088839.1 | - | 4, 5 | 425 | |||
BC023020.1 | - | 4, 5 | 392 | 5375425 |