Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190444.1 | NP_001177373.1 | 4 | 615 | ||
NM_146110.3 | NP_666222.2 | 4 | 612 | |||
GenBank mRNA | AK146470.1 | - | 4 | 555 | ||
AK151820.1 | - | 4 | 524 | |||
AK162750.1 | - | 4 | 540 | |||
AK167621.1 | - | 4 | 536 | |||
AK200115.1 | - | 3 | 256 | |||
BC031795.1 | - | 3 | 304 | 3965387 | ||
BC080766.1 | - | 2 | 1023 |