Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145999.2 | NP_666111.1 | 19 | 2442 | ||
GenBank mRNA | AK142183.1 | - | 14 | 2548 | ||
AY311390.1 | - | 19 | 2442 | |||
BC023163.1 | - | 9 | 1211 | |||
BC023927.1 | - | 5 | 1495 | |||
BC029777.1 | - | 19 | 2075 | 4950061 | ||
BC034062.1 | - | 19 | 2097 | 4207654 |