Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146003.2 | NP_666115.2 | 22 | 3686 | ||
GenBank mRNA | AK028461.1 | - | 10 | 1370 | ||
AK048485.1 | - | 17 | 3135 | |||
AK129219.1 | - | 18 | 3213 | |||
AK184730.1 | - | 3 | 127 | |||
BC028838.1 | - | 5 | 802 | |||
BC052718.1 | - | 22 | 3453 | |||
BC061480.1 | - | 22 | 3509 | 6509771 | ||
BC151128.1 | - | 23 | 3117 | 9088041 | ||
BC172171.1 | - | 23 | 3117 | 9054841 |