Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168679.1 | NP_001162150.1 | 9 | 1024 | ||
NM_001168680.1 | NP_001162151.1 | 9 | 1060 | |||
NM_146010.2 | NP_666122.1 | 8 | 1166 | |||
GenBank mRNA | AK077301.1 | - | 9 | 1074 | ||
AK085291.1 | - | 931 | ||||
BC025461.1 | - | 9 | 1032 | 5344986 | ||
BC027777.1 | - | 9 | 1035 | 5351814 | ||
BC029068.1 | - | 9 | 1056 | 4947664 | ||
BC029808.1 | - | 8 | 1165 | 4947699 | ||
BC034198.1 | - | 8 | 939 | 4986549 |