Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000666.2 | NP_000657.1 | 10 | 995 | ||
NM_001198895.1 | NP_001185824.1 | 10 | 990 | |||
NM_001198896.1 | NP_001185825.1 | 8 | 779 | |||
NM_001198898.1 | NP_001185827.1 | 9 | 890 | |||
GenBank mRNA | AK298274.1 | - | 10 | 744 | ||
BC000545.2 | - | 10 | 739 | 3162222 | ||
BC003023.1 | - | 10 | 732 | 2821665 | ||
BC014112.2 | - | 10 | 715 | 4543442 | ||
CD014023.1 | - | 8 | 620 | |||
CD014027.1 | - | 9 | 731 | |||
D14524.1 | - | 10 | 729 | |||
D16307.1 | - | 10 | 727 | |||
EU794586.1 | - | 10 | 715 | |||
KU177890.1 | - | 9 | 671 | |||
L07548.1 | - | 10 | 732 |
Gene Symbol: ABHD14A-ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316331.1 | NP_001303260.1 | 12 | 1196 | ||
GenBank mRNA | AK298086.1 | - | 12 | 1172 |