Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285785.1 | NP_001272714.1 | 16 | 2345 | ||
NM_146011.2 | NP_666123.2 | 15 | 1784 | |||
GenBank mRNA | AK049245.1 | - | 12 | 1565 | ||
AK080955.1 | - | 12 | 2974 | |||
AK083474.1 | - | 12 | 1726 | |||
AK089051.1 | - | 15 | 1736 | |||
AK167738.1 | - | 16 | 1922 | |||
BC021754.1 | - | 5 | 759 | |||
BC024535.1 | - | 15 | 1779 | 4987970 | ||
BC027374.1 | - | 15 | 1755 | 4947306 |