Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146013.1 | NP_666125.1 | 8 | 705 | ||
GenBank mRNA | AK028954.1 | - | 8 | 666 | ||
BC024073.1 | - | 1 | 40 | |||
BC026948.1 | - | 8 | 705 | 5136664 |