Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clptm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146047.2 | NP_666159.2 | 15, 16 | 1663 | ||
GenBank mRNA | AK032295.1 | - | 15, 16 | 1627 | ||
AK048357.1 | - | 15, 16 | 1560 | |||
AK156203.1 | - | 15, 16 | 1554 | |||
AK165939.1 | - | 15, 16 | 1524 | |||
AK181076.1 | - | 7, 8 | 385 | |||
AK187892.1 | - | 7, 8 | 369 | |||
BC026562.1 | - | 15, 16 | 1364 | 5028264 |