Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146052.4 | NP_666164.1 | 2 | 951 | ||
GenBank mRNA | AK043406.1 | - | 2 | 838 | ||
BC019794.1 | - | 2 | 713 | 4481142 |