Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146067.3 | NP_666179.2 | 4 | 1960 | ||
GenBank mRNA | AK029379.1 | - | 4 | 1885 | ||
AK032288.1 | - | 4 | 1923 | |||
AK049721.1 | - | 4 | 1961 | |||
AK080876.1 | - | 4 | 1918 | |||
AK133977.1 | - | 4 | 1954 | |||
BC025476.1 | - | 4 | 1881 | |||
BC034347.1 | - | 4 | 1602 | 2654874 |