Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163505.1 | NP_001156977.1 | 12 | 1541 | ||
NM_146091.4 | NP_666203.3 | 12 | 1443 | |||
GenBank mRNA | AK028842.1 | - | 12 | 1304 | ||
AK030660.1 | - | 12 | 1303 | |||
AK077752.1 | - | 12 | 1306 | |||
AK147720.1 | - | 12 | 1401 | |||
AK169476.1 | - | 12 | 1303 | |||
AK170550.1 | - | 12 | 2818 | |||
AK193433.1 | - | 2 | 183 | |||
BC002149.1 | - | 9 | 833 | |||
BC017138.1 | - | 12 | 1292 | 4485115 |