Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hibch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146108.1 | NP_666220.1 | 12 | 1041 | ||
GenBank mRNA | AK076038.1 | - | 12 | 1040 | ||
AK082965.1 | - | 12 | 1041 | |||
AK206848.1 | - | 5 | 352 | |||
AK214768.1 | - | 5 | 354 | |||
BC026437.1 | - | 12 | 1041 | 4238681 |