Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110214.1 | NP_001103684.1 | 7 | 610 | ||
NM_146114.3 | NP_666226.2 | 7 | 610 | |||
NM_175683.4 | NP_783614.1 | 7 | 610 | |||
GenBank mRNA | AF387731.1 | - | 7 | 592 | ||
AK037126.1 | - | 7 | 577 | |||
AK038788.1 | - | 7 | 540 | |||
AK052369.1 | - | 7 | 566 | |||
AK052490.1 | - | 7 | 549 | |||
AK088810.1 | - | 5 | 392 | |||
AK166324.1 | - | 7 | 553 | |||
BC108935.2 | - | 7 | 563 | 40039591 |