Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146122.3 | NP_666234.3 | 2, 3 | 336 | ||
GenBank mRNA | AK137530.1 | - | 2, 3 | 314 | ||
AK220407.1 | - | 2, 3 | 254 | |||
BC027786.1 | - | 2, 3 | 299 | 5346095 | ||
BQ444119.1 | - | 2, 3 | 174 | |||
DQ448594.1 | - | 2, 3 | 81 |