Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000689.4 | NP_000680.2 | 9 | 1248 | ||
GenBank mRNA | AF003341.1 | - | 9 | 931 | ||
AK000118.1 | - | 9 | 975 | |||
AK026641.1 | - | 9 | 975 | |||
AK293176.1 | - | 9 | 941 | |||
AK302168.1 | - | 8 | 759 | |||
AY390731.1 | - | 9 | 931 | |||
BC001505.2 | - | 9 | 956 | 2988388 | ||
BT006921.1 | - | 9 | 931 | |||
EU668318.1 | - | 9 | 975 | |||
EU794589.1 | - | 9 | 931 | |||
FJ224286.1 | - | 9 | 984 | |||
K03000.1 | - | 5 | 448 | |||
M26761.1 | - | 5 | 448 |