Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000690.3 | NP_000681.2 | 9 | 1091 | ||
NM_001204889.1 | NP_001191818.1 | 8 | 950 | |||
GenBank mRNA | AK223373.1 | - | 9 | 1033 | ||
AK314856.1 | - | 9 | 1012 | |||
AY621070.1 | - | 9 | 1056 | |||
BC002967.1 | - | 9 | 1041 | 3543343 | ||
BC071839.1 | - | 9 | 1000 | 5477768 | ||
CR456991.1 | - | 9 | 995 | |||
K03001.1 | - | 6 | 641 | |||
M26760.1 | - | 6 | 641 | |||
M54931.1 | - | 4 | 356 | |||
X05409.1 | - | 9 | 1028 | |||
Y00109.1 | - | 9 | 1019 |