Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301414.1 | NP_001288343.1 | 2 | 604 | ||
NM_146144.4 | NP_666256.2 | 2 | 585 | |||
GenBank mRNA | AK080882.1 | - | 2 | 586 | ||
BC018179.1 | - | 2 | 465 | 3585191 | ||
BC020007.1 | - | 2 | 437 | 3980469 | ||
BY713495.1 | - | 2 | 473 |