Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000694.3 | NP_000685.1 | 10 | 2281 | ||
NM_001030010.2 | NP_001025181.1 | 9 | 2170 | |||
NM_001161473.2 | NP_001154945.1 | 10 | 2339 | |||
NM_001290058.1 | NP_001276987.1 | 9 | 1930 | |||
NM_001290059.1 | NP_001276988.1 | 9 | 2199 | |||
GenBank mRNA | AB209651.1 | - | 8 | 4436 | ||
BC013584.2 | - | 10 | 2234 | 3878124 | ||
BC014168.2 | - | 4 | 1269 | |||
U10868.1 | - | 10 | 2242 |