Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 550 | ||
NM_146184.4 | NP_666296.1 | 1 | 942 | |||
GenBank mRNA | AK042388.1 | - | 1 | 925 | ||
AK154220.1 | - | 537 | ||||
AK154901.1 | - | 2 | 506 | |||
BC025206.1 | - | 2 | 536 | 5037295 |