Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146191.3 | NP_666303.3 | 32 | 5599 | ||
GenBank mRNA | AK040153.1 | - | 12 | 2281 | ||
AK076432.1 | - | 5 | 926 | |||
AK081849.1 | - | 2 | 592 | |||
AK129441.1 | - | 29 | 5012 | |||
AK147469.1 | - | 32 | 5585 | |||
AK154395.1 | - | 6 | 1118 | |||
BC027199.1 | - | 2 | 467 | |||
BC066159.1 | - | 1 | 121 | |||
BC080819.1 | - | 1 | 226 |