Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000701.7 | NP_000692.2 | 9 | 1405 | ||
NM_001160233.1 | NP_001153705.1 | 9 | 1429 | |||
NM_001160234.1 | NP_001153706.1 | 9 | 1238 | |||
XM_017001360.1 | XP_016856849.1 | 10 | 4624 | |||
XM_017001361.1 | XP_016856850.1 | 10 | 3966 | |||
GenBank mRNA | AK295095.1 | - | 9 | 1428 | ||
AK296362.1 | - | 9 | 1238 | |||
AK296384.1 | - | 9 | 1364 | |||
AK308418.1 | - | 9 | 1170 | |||
AK314777.1 | - | 9 | 1250 | |||
BC003077.2 | - | 9 | 1318 | 3507685 | ||
BC050359.1 | - | 9 | 1316 | 6065209 | ||
J03007.1 | - | 4 | 475 | |||
U16798.1 | - | 8 | 1066 |