Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206367.1 | NP_001193296.1 | 5 | 574 | ||
NM_001206368.1 | NP_001193297.1 | 4 | 490 | |||
NM_001206369.1 | NP_001193298.1 | 4 | 535 | |||
NM_146120.4 | NP_666232.2 | 4 | 597 | |||
GenBank mRNA | AK076156.1 | - | 4 | 537 | ||
AK089934.1 | - | 5 | 529 | |||
AK135632.1 | - | 3 | 310 | |||
AK143664.1 | - | 4 | 564 | |||
AK146969.1 | - | 5 | 491 | |||
AK148957.1 | - | 5 | 554 | |||
AK150359.1 | - | 5 | 497 | |||
AK151684.1 | - | 5 | 516 | |||
AK153029.1 | - | 5 | 494 | |||
AK166831.1 | - | 4 | 510 | |||
AK166932.1 | - | 5 | 488 | |||
AK167918.1 | - | 5 | 504 | |||
AK168461.1 | - | 5 | 507 | |||
AK168532.1 | - | 5 | 508 | |||
AK168707.1 | - | 4 | 443 | |||
AK168725.1 | - | 5 | 491 | |||
AK168829.1 | - | 4 | 542 | |||
AK169041.1 | - | 5 | 494 | |||
AK169670.1 | - | 5 | 519 | |||
BC023143.1 | - | 4 | 535 | 3711602 | ||
BC060377.1 | - | 5 | 522 | 4983664 | ||
J04953.1 | - | 3 | 384 |