Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C87436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243741.1 | NP_001230670.1 | 3 | 1671 | ||
NM_001243742.1 | NP_001230671.1 | 3 | 1207 | |||
NM_146170.4 | NP_666282.3 | 3 | 1148 | |||
GenBank mRNA | AK047127.1 | - | 3 | 1149 | ||
AK077101.1 | - | 974 | ||||
AK077503.1 | - | 3 | 1558 | |||
AK170524.1 | - | 4 | 1039 | |||
AK171511.1 | - | 4 | 1039 | |||
AK178467.1 | - | 1 | 137 | |||
BC025632.1 | - | 3 | 1115 | 5321669 | ||
BC026926.1 | - | 3 | 1218 | 3586097 | ||
BC031913.1 | - | 3 | 1161 | 5376246 |