Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146175.3 | NP_666287.3 | 2 | 316 | ||
GenBank mRNA | AK030516.1 | - | 2 | 225 | ||
AK081854.1 | - | 2 | 215 | |||
AK088531.1 | - | 2 | 315 | |||
AK148038.1 | - | 2 | 302 | |||
AK188102.1 | - | 2 | 134 | |||
BC025581.1 | - | 2 | 218 | 5325029 |