Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146207.2 | NP_666319.2 | 6 | 675 | ||
GenBank mRNA | AK144802.1 | - | 6 | 657 | ||
AK150237.1 | - | 6 | 654 | |||
AK166251.1 | - | 6 | 657 | |||
AK168215.1 | - | 6 | 657 | |||
AK170722.1 | - | 6 | 657 | |||
AK183546.1 | - | 1 | 25 | |||
AK215945.1 | - | 1 | 37 | |||
AK217754.1 | - | 1 | 37 | |||
BC010211.1 | - | 6 | 653 | 3969515 | ||
BC024113.1 | - | 2 | 147 | 5097944 | ||
BC027006.1 | - | 1 | 70 |