Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146059.1 | NP_001139531.1 | 13 | 1403 | ||
NM_001146060.1 | NP_001139532.1 | 13 | 1483 | |||
NM_146228.4 | NP_666340.3 | 13 | 1400 | |||
GenBank mRNA | AB107016.1 | - | 13 | 1400 | ||
AF481142.1 | - | 12 | 1296 | |||
AK030815.1 | - | 6 | 537 | |||
AK053076.1 | - | 13 | 1373 | |||
AK131149.1 | - | 13 | 1402 | |||
AK155451.1 | - | 13 | 1469 | |||
BC117928.1 | - | 12 | 1353 | 40097635 | ||
BC117929.1 | - | 12 | 1350 | 40097636 |