Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146229.2 | NP_666341.1 | 3 | 332 | ||
GenBank mRNA | AK082950.1 | - | 3 | 373 | ||
AK159702.1 | - | 374 | ||||
AK180961.1 | - | 1 | 75 | |||
BC023347.1 | - | 3 | 332 | 5040044 |