Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubb4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146116.2 | NP_666228.1 | 4 | 486 | ||
GenBank mRNA | AK006924.1 | - | 3 | 250 | ||
AK015901.1 | - | 4 | 439 | |||
AK076895.1 | - | 4 | 443 | |||
AK146587.1 | - | 4 | 442 | |||
AK167022.1 | - | 4 | 441 | |||
BC005547.1 | - | 4 | 418 | 3585500 | ||
BC022919.1 | - | 4 | 425 | 4221189 | ||
BC083319.1 | - | 4 | 428 | 5699674 |