Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164357.1 | NP_001157829.1 | 10 | 2490 | ||
NM_001164358.1 | NP_001157830.1 | 11 | 2745 | |||
NM_001290558.1 | NP_001277487.1 | 10 | 2359 | |||
NM_146118.3 | NP_666230.2 | 11 | 2526 | |||
GenBank mRNA | AK049392.1 | - | 10 | 2466 | ||
AK082756.1 | - | 10 | 2447 | |||
AK122568.2 | - | 11 | 2711 | |||
AK132204.1 | - | 10 | 2444 | |||
BC004720.1 | - | 3 | 1216 | |||
BC019978.1 | - | 10 | 2298 | 4239441 | ||
BC022114.1 | - | 10 | 2296 | 5098924 | ||
BC037109.1 | - | 11 | 2486 | 5368306 | ||
BC066998.1 | - | 11 | 2713 | 5703622 |