Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam129b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146119.2 | NP_666231.1 | 14 | 2993 | ||
GenBank mRNA | AK033735.1 | - | 14 | 2960 | ||
AK046583.1 | - | 14 | 2890 | |||
AK051396.1 | - | 14 | 2932 | |||
AK146953.1 | - | 14 | 2978 | |||
AK149818.1 | - | 14 | 2903 | |||
AK149888.1 | - | 14 | 2903 | |||
AK150642.1 | - | 14 | 2931 | |||
AK152207.1 | - | 14 | 2952 | |||
AK153489.1 | - | 14 | 2936 | |||
AK170512.1 | - | 14 | 2933 | |||
BC024639.1 | - | 6 | 1726 | |||
BC027843.1 | - | 14 | 2913 | 5343013 |