Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146129.3 | NP_666241.1 | 16, 17 | 2200 | ||
GenBank mRNA | AK041830.1 | - | 206 | |||
AK089358.1 | - | 16, 17 | 2201 | |||
BC029713.1 | - | 3, 4 | 298 | |||
BC031431.1 | - | 16, 17 | 2169 | 4241283 |