Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146136.1 | NP_666248.1 | 9 | 1828 | ||
GenBank mRNA | AK045797.1 | - | 9 | 1814 | ||
AK049873.1 | - | 9 | 1891 | |||
AK085492.1 | - | 9 | 1825 | |||
AK136837.1 | - | 9 | 1827 | |||
BC026596.1 | - | 9 | 1828 | 4975010 |