Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290725.1 | NP_001277654.1 | 7 | 1918 | ||
NM_146154.3 | NP_666266.1 | 7 | 1921 | |||
GenBank mRNA | AK032022.1 | - | 5 | 1731 | ||
AK198369.1 | - | 1 | 254 | |||
AX775983.1 | - | 7 | 1887 | |||
BC025479.1 | - | 7 | 1883 | 5354224 | ||
BQ749177.1 | - | 1 | 298 |