Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146155.3 | NP_666267.3 | 6, 7 | 5473 | ||
GenBank mRNA | AK080410.1 | - | 1, 2 | 455 | ||
AK149347.1 | - | 7, 8 | 3500 | |||
BC019130.1 | - | 1, 2 | 1072 | |||
BC042621.1 | - | 6, 7 | 5462 | 4911638 | ||
BC060231.1 | - | 5, 6 | 5441 | 6852595 |