Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000757.5 | NP_000748.3 | 4 | 796 | ||
NM_172210.2 | NP_757349.1 | 4 | 796 | |||
NM_172211.3 | NP_757350.1 | 4 | 796 | |||
NM_172212.2 | NP_757351.1 | 4 | 796 | |||
XM_017000369.1 | XP_016855858.1 | 4 | 471 | |||
GenBank mRNA | AB208952.1 | - | 4 | 545 | ||
AK291660.1 | - | 4 | 558 | |||
AK300401.1 | - | 2 | 178 | |||
AK311440.1 | - | 1 | 88 | |||
BC021117.1 | - | 4 | 572 | 4586312 | ||
JF803735.1 | - | 3 | 287 | |||
M27087.1 | - | 4 | 507 | |||
M37435.1 | - | 3 | 488 | |||
M64592.1 | - | 4 | 383 | |||
M76453.1 | - | 3 | 347 | |||
U22386.1 | - | 4 | 427 | |||
X05825.1 | - | 3 | 331 |