Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146205.2 | NP_666317.2 | 6 | 3608 | ||
GenBank mRNA | AK030000.1 | - | 6 | 2465 | ||
AK131111.1 | - | 6 | 3608 | |||
BC025196.1 | - | 3 | 1079 | |||
BC032200.1 | - | 6 | 2953 | 5342166 | ||
BC051105.1 | - | 2 | 868 | |||
BC089363.1 | - | 6 | 2849 | 5720275 |