Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vac14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146216.2 | NP_666328.2 | 6, 7 | 950 | ||
GenBank mRNA | AK031952.1 | - | 6, 7 | 985 | ||
AK043319.1 | - | 6, 7 | 982 | |||
AK046826.1 | - | 6, 7 | 922 | |||
AK084520.1 | - | 6, 7 | 1001 | |||
AK140191.1 | - | 6, 7 | 977 | |||
AK171577.1 | - | 6, 7 | 990 | |||
AK171883.1 | - | 6, 7 | 978 | |||
BC052199.1 | - | 6, 7 | 950 | 6512678 |