Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfwd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146218.4 | NP_666330.2 | 5 | 903 | ||
GenBank mRNA | AK137499.1 | - | 5 | 883 | ||
AK137698.1 | - | 5 | 866 | |||
AK139032.1 | - | 5 | 884 | |||
AK146451.1 | - | 5 | 873 | |||
BC018533.1 | - | 5 | 872 | 2651526 | ||
BC023694.1 | - | 5 | 908 | 5344528 | ||
BC096602.1 | - | 5 | 866 | 4954068 |