Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 5 | 1037 | ||
NM_001099773.1 | NP_001093243.1 | 5 | 1111 | |||
GenBank mRNA | AK056794.1 | - | 5 | 1106 | ||
AK292300.1 | - | 5 | 943 | |||
AK292301.1 | - | 4 | 901 | |||
AK293204.1 | - | 4 | 743 | |||
AK293211.1 | - | 6 | 903 | |||
AK300178.1 | - | 5 | 853 | |||
BC032329.1 | - | 5 | 926 | 5165263 | ||
KU178011.1 | - | 5 | 882 | |||
M14565.1 | - | 5 | 926 | |||
M28253.1 | - | 1 | 36 |