Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146236.1 | NP_666348.1 | 2 | 167 | ||
GenBank mRNA | BC011290.1 | - | 2 | 167 | 4010035 | |
BC083157.1 | - | 2 | 201 | 5376199 |