Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cchcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146248.2 | NP_666360.2 | 3 | 362 | ||
GenBank mRNA | AK159780.1 | - | 3 | 289 | ||
BC018431.1 | - | 3 | 362 | |||
BC031416.1 | - | 4 | 451 | 4012768 | ||
BM947393.1 | - | 3 | 274 |