Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146238.3 | NP_666350.2 | 3 | 460 | ||
GenBank mRNA | AK037580.1 | - | 3 | 356 | ||
AK047511.1 | - | 4 | 555 | |||
AK053774.1 | - | 3 | 424 | |||
AK160583.1 | - | 3 | 461 | |||
BC023488.1 | - | 4 | 450 | 5010787 |